missing translation for 'onlineSavingsMsg'
Learn More

Bis[2-(di-tert-butylphosphino)ethyl]amine, 10% w/w soln. in THF

CAS: 944710-34-3 | C20H45NP2 | 361.54 g/mol

Supplier:  Thermo Scientific Chemicals H6049306

 View more versions of this product

Catalog No. AAH6049306


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
This compound is involved in the transition metal-catalyzed addition of amines to acrylic acid derivatives.

Solubility
Insoluble in water.

Notes
Air Sensitive. Incompatible with oxidizing agents.
Specifications

Specifications

Bis[2-(di-tert-butylphosphino)ethyl]amine
0.911
C20H45NP2
([(CH3)3C]2PCH2CH2)2NH
5 g
Air Sensitive
Insoluble in water.
CC(C)(C)P(CCNCCP(C(C)(C)C)C(C)(C)C)C(C)(C)C
361.54
361.53
944710-34-3
Unpleasant
1.418
MFCD17014018
UN1993
bis 2-di-t-butylphosphino ethyl amine, bis 2-di-tert-butylphosphino ethyl amine, bis 2-di-tert-butylphosphanyl ethyl amine, bis 2-di-tert-butylphosphino ethyl amine w/w soln. in thf, bis 2-di-tert-butylphosphino ethyl amine solution, 10 wt. % in thf
MJYKYLNPIGDVEF-UHFFFAOYSA-N
2-ditert-butylphosphanyl-N-(2-ditert-butylphosphanylethyl)ethanamine
51346231
10% w/w soln. in THF

Chemical Identifiers

944710-34-3
361.54
MJYKYLNPIGDVEF-UHFFFAOYSA-N
51346231
CC(C)(C)P(CCNCCP(C(C)(C)C)C(C)(C)C)C(C)(C)C
C20H45NP2
MFCD17014018
bis 2-di-t-butylphosphino ethyl amine, bis 2-di-tert-butylphosphino ethyl amine, bis 2-di-tert-butylphosphanyl ethyl amine, bis 2-di-tert-butylphosphino ethyl amine w/w soln. in thf, bis 2-di-tert-butylphosphino ethyl amine solution, 10 wt. % in thf
2-ditert-butylphosphanyl-N-(2-ditert-butylphosphanylethyl)ethanamine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H351-H315-H319-H335
Highly flammable liquid and vapor.
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P201-P202-P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P310-P330-P332+P313-P363-P370+P378q-P501c

H225-H302-H315-H318-H335-H336-H351

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only