missing translation for 'onlineSavingsMsg'
Learn More

Bis[2-(di-tert-butylphosphino)ethyl]amine, 10% w/w soln. in THF

Catalog No. AAH6049303
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6049303 1 g
AAH6049306 5 g
AAH6049314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6049303 Supplier Thermo Scientific Chemicals Supplier No. H6049303

CAS: 944710-34-3 | C20H45NP2 | 361.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
This compound is involved in the transition metal-catalyzed addition of amines to acrylic acid derivatives.

Solubility
Insoluble in water.

Notes
Air Sensitive. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 944710-34-3
Molecular Formula C20H45NP2
Molecular Weight (g/mol) 361.54
MDL Number MFCD17014018
InChI Key MJYKYLNPIGDVEF-UHFFFAOYSA-N
Synonym bis 2-di-t-butylphosphino ethyl amine, bis 2-di-tert-butylphosphino ethyl amine, bis 2-di-tert-butylphosphanyl ethyl amine, bis 2-di-tert-butylphosphino ethyl amine w/w soln. in thf, bis 2-di-tert-butylphosphino ethyl amine solution, 10 wt. % in thf
PubChem CID 51346231
IUPAC Name 2-ditert-butylphosphanyl-N-(2-ditert-butylphosphanylethyl)ethanamine
SMILES CC(C)(C)P(CCNCCP(C(C)(C)C)C(C)(C)C)C(C)(C)C

Specifications

Density 0.911
Odor Unpleasant
Refractive Index 1.418
Linear Formula ([(CH3)3C]2PCH2CH2)2NH
Quantity 1 g
UN Number UN1993
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 361.53
Concentration or Composition (by Analyte or Components) 10% w/w soln. in THF
Chemical Name or Material Bis[2-(di-tert-butylphosphino)ethyl]amine
Show More Show Less
Hazard Category H225-H302-H315-H318-H335-H336-H351
Hazard Statement GHS H Statement
H225-H351-H315-H319-H335
Highly flammable liquid and vapor.
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P310-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only