missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-cyanoethyl) ether, 98%

Catalog No. AAA1100509
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1100509 10 g
AAA1100518 50 g
AAA1100530 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1100509 Supplier Thermo Scientific Chemicals Supplier No. A1100509
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1656-48-0 | C6H8N2O | 124.143 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1656-48-0
Molecular Formula C6H8N2O
Molecular Weight (g/mol) 124.143
MDL Number MFCD00001960
InChI Key BCGCCTGNWPKXJL-UHFFFAOYSA-N
Synonym bis 2-cyanoethyl ether, 2-cyanoethyl ether, 3,3'-oxydipropionitrile, propanenitrile, 3,3'-oxybis, ether, bis 2-cyanoethyl, 2,2'-dicyanodiethyl ether, propionitrile, 3,3'-oxydi, 3-2-cyanoethoxy propanenitrile, beta,beta'-dicyanodiethyl ether, unii-uf1uoo2pho
PubChem CID 15452
IUPAC Name 3-(2-cyanoethoxy)propanenitrile
SMILES C(COCCC#N)C#N

Specifications

Density 1.043
Boiling Point 119°C to 121°C (1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.44
Quantity 10 g
Formula Weight 124.11
Percent Purity 98%
Chemical Name or Material Bis (2-cyanoethyl) ether
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 216-750-9
RTECSNumber UG4200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only