missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(2-chloroethyl) Ether 99.0+%, TCI America™
$57.79 - $113.71
Chemical Identifiers
| CAS | 111-44-4 |
|---|---|
| Molecular Formula | C4H8Cl2O |
| Molecular Weight (g/mol) | 143.01 |
| MDL Number | MFCD00000975 |
| InChI Key | ZNSMNVMLTJELDZ-UHFFFAOYSA-N |
| Synonym | 2-chloroethyl ether, 2,2'-dichlorodiethyl ether, bis 2-chloroethyl ether, chlorex, chloroethyl ether, dcee, dichloroether, khloreks, dichloroethyl ether |
| PubChem CID | 8115 |
| ChEBI | CHEBI:34573 |
| IUPAC Name | 1-chloro-2-(2-chloroethoxy)ethane |
| SMILES | ClCCOCCCl |
Chemical Identifiers
| 111-44-4 | |
| 143.01 | |
| ZNSMNVMLTJELDZ-UHFFFAOYSA-N | |
| 8115 | |
| 1-chloro-2-(2-chloroethoxy)ethane |
| C4H8Cl2O | |
| MFCD00000975 | |
| 2-chloroethyl ether, 2,2'-dichlorodiethyl ether, bis 2-chloroethyl ether, chlorex, chloroethyl ether, dcee, dichloroether, khloreks, dichloroethyl ether | |
| CHEBI:34573 | |
| ClCCOCCCl |
Specifications
| 111-44-4 | |
| Yellow | |
| C4H8Cl2O | |
| 25 g | |
| 2-chloroethyl ether, 2,2'-dichlorodiethyl ether, bis 2-chloroethyl ether, chlorex, chloroethyl ether, dcee, dichloroether, khloreks, dichloroethyl ether | |
| ClCCOCCCl | |
| 143.01 | |
| CHEBI:34573 | |
| ≥99.0% (GC) | |
| Bis(2-chloroethyl) Ether |
| -47°C | |
| 178°C | |
| MFCD00000975 | |
| 1916 | |
| ZNSMNVMLTJELDZ-UHFFFAOYSA-N | |
| 1-chloro-2-(2-chloroethoxy)ethane | |
| 8115 | |
| 143.01 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0382
RTECSNumber : KN0875000
TSCA : Yes