missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-chloroethyl) Ether 99.0+%, TCI America™

Catalog No. B0472500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0472500G 500 g
B047225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0472500G Supplier TCI America Supplier No. B0472500G

Chemical Identifiers

CAS 111-44-4
Molecular Formula C4H8Cl2O
Molecular Weight (g/mol) 143.01
MDL Number MFCD00000975
InChI Key ZNSMNVMLTJELDZ-UHFFFAOYSA-N
Synonym 2-chloroethyl ether, 2,2'-dichlorodiethyl ether, bis 2-chloroethyl ether, chlorex, chloroethyl ether, dcee, dichloroether, khloreks, dichloroethyl ether
PubChem CID 8115
ChEBI CHEBI:34573
IUPAC Name 1-chloro-2-(2-chloroethoxy)ethane
SMILES ClCCOCCCl

Specifications

Melting Point -47°C
Color Yellow
Boiling Point 178°C
Quantity 500 g
UN Number 1916
Formula Weight 143.01
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Bis(2-chloroethyl) Ether
EINECSNumber (2)-0382
RTECSNumber KN0875000
TSCA Yes