missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-chloroethyl) ether, 99%

CAS: 111-44-4 | C4H8Cl2O | 143.01 g/mol

Supplier:  Thermo Scientific Chemicals A142340B

 View more versions of this product

Catalog No. AAA142340B


Only null left
Explore available promotions

Description

Description

Bis(2-chloroethyl) ether is used as a solvent and soil fumigant. It may be used in the preparation of macrocyclic polyethers, dibenzo-18-crown-6 polyether and dicyclohexyl-18-crown-6 polyether.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(2-chloroethyl) ether is used as a solvent and soil fumigant. It may be used in the preparation of macrocyclic polyethers, dibenzo-18-crown-6 polyether and dicyclohexyl-18-crown-6 polyether.

Solubility
Soluble in water. (1.1 g/L) at 20°C.

Notes
Special handling precautions required. View MSDS prior to purchase. MSDS are available online at www.alfa.com
Specifications

Specifications

Bis(2-chloroethyl) ether
-50°C
178°C
Pungent
1.457
MFCD00000975
UN1916
14,3066
Soluble in water. (1.1g/L) at 20°C.
ClCCOCCCl
143.01
CHEBI:34573
99%
111-44-4
1.219
55°C (131°F)
C4H8Cl2O
ClCH2CH2OCH2CH2Cl
1000 g
605317
2-chloroethyl ether, 2,2'-dichlorodiethyl ether, bis 2-chloroethyl ether, chlorex, chloroethyl ether, dcee, dichloroether, khloreks, dichloroethyl ether
ZNSMNVMLTJELDZ-UHFFFAOYSA-N
1-chloro-2-(2-chloroethoxy)ethane
8115
143.01

Chemical Identifiers

111-44-4
143.01
ZNSMNVMLTJELDZ-UHFFFAOYSA-N
8115
1-chloro-2-(2-chloroethoxy)ethane
C4H8Cl2O
MFCD00000975
2-chloroethyl ether, 2,2'-dichlorodiethyl ether, bis 2-chloroethyl ether, chlorex, chloroethyl ether, dcee, dichloroether, khloreks, dichloroethyl ether
CHEBI:34573
ClCCOCCCl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330-H226-H351
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Flammable liquid and vapour.
Suspected of causing cancer.

P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P280-P284-P301+P310-P303+P361+P353-P304+P340-P310-P330-P363-P370+P378q-P501c

H226-H300+H310+H330

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: 2,2'-DICHLORODIETHYL ETHER

EINECSNumber : 203-870-1

RTECSNumber : KN0875000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only