missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-chloroethyl) ether, 99%

Catalog No. AAA1423430
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1423430 250 g
AAA1423418 50 g
AAA142340B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1423430 Supplier Thermo Scientific Chemicals Supplier No. A1423430
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 111-44-4 | C4H8Cl2O | 143.01 g/mol

Bis(2-chloroethyl) ether is used as a solvent and soil fumigant. It may be used in the preparation of macrocyclic polyethers, dibenzo-18-crown-6 polyether and dicyclohexyl-18-crown-6 polyether.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(2-chloroethyl) ether is used as a solvent and soil fumigant. It may be used in the preparation of macrocyclic polyethers, dibenzo-18-crown-6 polyether and dicyclohexyl-18-crown-6 polyether.

Solubility
Soluble in water. (1.1 g/L) at 20°C.

Notes
Special handling precautions required. View MSDS prior to purchase. MSDS are available online at www.alfa.com

Chemical Identifiers

CAS 111-44-4
Molecular Formula C4H8Cl2O
Molecular Weight (g/mol) 143.01
MDL Number MFCD00000975
InChI Key ZNSMNVMLTJELDZ-UHFFFAOYSA-N
Synonym 2-chloroethyl ether, 2,2'-dichlorodiethyl ether, bis 2-chloroethyl ether, chlorex, chloroethyl ether, dcee, dichloroether, khloreks, dichloroethyl ether
PubChem CID 8115
ChEBI CHEBI:34573
IUPAC Name 1-chloro-2-(2-chloroethoxy)ethane
SMILES ClCCOCCCl

Specifications

Melting Point -50°C
Density 1.219
Boiling Point 178°C
Flash Point 55°C (131°F)
Odor Pungent
Refractive Index 1.457
Linear Formula ClCH2CH2OCH2CH2Cl
Quantity 250 g
UN Number UN1916
Beilstein 605317
Merck Index 14,3066
Solubility Information Soluble in water. (1.1g/L) at 20°C.
Formula Weight 143.01
Percent Purity 99%
Chemical Name or Material Bis(2-chloroethyl) ether
Show More Show Less
Hazard Category H226-H300+H310+H330
Hazard Statement GHS H Statement
H300-H310-H330-H226-H351
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Flammable liquid and vapour.
Suspected of causing cancer.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P280-P284-P301+P310-P303+P361+P353-P304+P340-P310-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: 2,2'-DICHLORODIETHYL ETHER
EINECSNumber 203-870-1
RTECSNumber KN0875000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only