missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(2-butoxyethyl) Adipate 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | -34°C |
| Color | Yellow |
| Boiling Point | 217°C |
| Quantity | 500 g |
| Formula Weight | 346.46 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Bis(2-butoxyethyl) Adipate |
Safety and Handling
| EINECSNumber | (2)-0883&(2)-0884 |
| RTECSNumber | AU8450000 |
| TSCA | Yes |