missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-butoxyethyl) Adipate 97.0+%, TCI America™

Catalog No. B271625G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B271625G 25 g
B2716500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B271625G Supplier TCI America Supplier No. B271625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 141-18-4
Molecular Formula C18H34O6
Molecular Weight (g/mol) 346.46
MDL Number MFCD00053796
InChI Key IHTSDBYPAZEUOP-UHFFFAOYSA-N
Synonym Adipic Acid Bis(2-butoxyethyl) Ester, Adipic Acid Di(2-butoxyethyl) Ester, Di(2-butoxyethyl) Adipate
PubChem CID 8837
IUPAC Name 1,6-bis(2-butoxyethyl) hexanedioate
SMILES CCCCOCCOC(=O)CCCCC(=O)OCCOCCCC

Specifications

Melting Point -34°C
Color Yellow
Boiling Point 217°C
Quantity 25 g
Formula Weight 346.46
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Bis(2-butoxyethyl) Adipate
EINECSNumber (2)-0883&(2)-0884
RTECSNumber AU8450000
TSCA Yes