missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-bromoethyl) Ether 98.0+%, TCI America™

Catalog No. B375225G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B375225G 25 g
B37525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B375225G Supplier TCI America Supplier No. B375225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5414-19-7
Molecular Formula C4H8Br2O
Molecular Weight (g/mol) 231.915
MDL Number MFCD00039196
InChI Key FOZVXADQAHVUSV-UHFFFAOYSA-N
Synonym bis 2-bromoethyl ether, 1-bromo-2-2-bromoethoxy ethane, 2-bromoethyl ether, 2,2'-dibromodiethyl ether, ethane, 1,1'-oxybis 2-bromo, ether, bis 2-bromoethyl, beta,beta'-dibromodiethyl ether, 2,2-dibromodiethyl ether, 2-bromoethylether
PubChem CID 21521
IUPAC Name 1-bromo-2-(2-bromoethoxy)ethane
SMILES C(CBr)OCCBr

Specifications

Color Yellow
Boiling Point 45°C
Quantity 25 g
Formula Weight 231.92
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Bis(2-bromoethyl) Ether
EINECSNumber (2)-3529
TSCA Yes