missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-bromoethyl) ether, 95%

Catalog No. AAL1151014
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1151014 25 g
AAL1151006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1151014 Supplier Thermo Scientific Chemicals Supplier No. L1151014

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5414-19-7 | C4H8Br2O | 231.915 g/mol

Bis(2-bromoethyl) ether is used dfor synthesis of heterocyclic and crown ether. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(2-bromoethyl) ether is used dfor synthesis of heterocyclic and crown ether. It is also used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases.

Chemical Identifiers

CAS 5414-19-7
Molecular Formula C4H8Br2O
Molecular Weight (g/mol) 231.915
MDL Number MFCD00039196
InChI Key FOZVXADQAHVUSV-UHFFFAOYSA-N
Synonym bis 2-bromoethyl ether, 1-bromo-2-2-bromoethoxy ethane, 2-bromoethyl ether, 2,2'-dibromodiethyl ether, ethane, 1,1'-oxybis 2-bromo, ether, bis 2-bromoethyl, beta,beta'-dibromodiethyl ether, 2,2-dibromodiethyl ether, 2-bromoethylether
PubChem CID 21521
IUPAC Name 1-bromo-2-(2-bromoethoxy)ethane
SMILES C(CBr)OCCBr

Specifications

Density 1.845
Boiling Point 92°C to 93°C (12 mmHg)
Flash Point 85°C (185°F)
Refractive Index 1.514
Quantity 25 g
UN Number UN2810
Beilstein 1098335
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 231.93
Percent Purity 96%
Chemical Name or Material Bis(2-bromoethyl) ether
Show More Show Less
Hazard Category H227-H315-H318-H335
Hazard Statement GHS H Statement
H300-H310-H330-H318-H351-H227-H315-H335
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes serious eye damage.
Suspected of causing cancer.
Combustible liquid.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 226-504-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only