missing translation for 'onlineSavingsMsg'
Learn More

Bis(2,4-pentanedionato)zinc(II) 96.0+%, TCI America™

Catalog No. Z0002500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Z0002500G 500 g
Z000225G 25 g
Z0002100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. Z0002500G Supplier TCI America Supplier No. Z0002500G
Only null left

Chemical Identifiers

CAS 14024-63-6
Molecular Formula C10H14O4Zn
Molecular Weight (g/mol) 263.60
MDL Number MFCD00000035
InChI Key CYDXJXDAFPJUQE-FDGPNNRMSA-L
Synonym zinc acetylacetonate hydrate, zinc 2,4-pentanedionate, hydrate, zinc 2+ bis 2z-4-oxopent-2-en-2-olate hydrate
PubChem CID 131675103
IUPAC Name zinc(2+) bis((2Z)-4-oxopent-2-en-2-olate)
SMILES [Zn++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Color White
Physical Form Crystalline Powder
Quantity 500 g
Formula Weight 263.60
Percent Purity ≥96.0% (T)
Chemical Name or Material Bis(2,4-pentanedionato)zinc(II)
EINECSNumber (2)-2181
RTECSNumber ZH0917500
TSCA Yes