missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(2,4-pentanedionato)zinc(II) 96.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Physical Form | Crystalline Powder |
| Quantity | 25 g |
| Formula Weight | 263.60 |
| Percent Purity | ≥96.0% (T) |
| Chemical Name or Material | Bis(2,4-pentanedionato)zinc(II) |
Safety and Handling
| EINECSNumber | (2)-2181 |
| RTECSNumber | ZH0917500 |
| TSCA | Yes |