missing translation for 'onlineSavingsMsg'
Learn More

Bis(2,4-pentanedionato)vanadium(IV) Oxide 95.0+%, TCI America™

Catalog No. V001625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V001625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. V001625G Supplier TCI America Supplier No. V001625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3153-26-2
Molecular Formula C10H14O5V
Molecular Weight (g/mol) 265.16
MDL Number MFCD00000032
InChI Key JFHJZWAQYMGNBE-SUKNRPLKSA-L
Synonym vanadyl acetylacetonate, bis 2,4-pentanedionato vanadium iv oxide
PubChem CID 131674261
IUPAC Name oxovanadiumbis(ylium) bis((2Z)-4-oxopent-2-en-2-olate)
SMILES O=[V++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Color Blue
Quantity 25 g
Formula Weight 265.16
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(2,4-pentanedionato)vanadium(IV) Oxide
EINECSNumber (2)-1869
TSCA Yes