missing translation for 'onlineSavingsMsg'
Learn More

Bis(2,4-pentanedionato)platinum(II), TCI America™

Catalog No. B31151G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31151G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B31151G Supplier TCI America Supplier No. B31151G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 15170-57-7
Molecular Formula C10H14O4Pt
Molecular Weight (g/mol) 393.30
MDL Number MFCD00000028
InChI Key KLFRPGNCEJNEKU-FDGPNNRMSA-L
Synonym platinum ii acetylacetonate, pt acac 2, platinum bis acetylacetonate, acetylacetone platinum ii salt, platinum 2,4-pentanedionate, 2,4-pentanedione platinum ii derivative, bis acetylacetonato platinum, bis acetylacetonato platinum ii, 2,4-pentanedione, platinum ii
PubChem CID 10960186
IUPAC Name platinum(2+) bis((2Z)-4-oxopent-2-en-2-olate)
SMILES [Pt++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point 250°C
Color Yellow
Quantity 1 g
Formula Weight 393.30
Physical Form Crystalline Powder
Chemical Name or Material Bis(2,4-pentanedionato)platinum(II)
EINECSNumber (2)-2181
TSCA Yes