missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(2,4-pentanedionato)palladium(II) 98.0+%, TCI America™
$348.57 - $1735.43
Chemical Identifiers
| CAS | 14024-61-4 |
|---|---|
| Molecular Formula | C10H14O4Pd |
| Molecular Weight (g/mol) | 304.64 |
| MDL Number | MFCD00000025 MFCD00000025 |
| InChI Key | JKDRQYIYVJVOPF-FDGPNNRMSA-L |
| Synonym | palladium diacetonate, acetylacetone palladium ii salt, bis 2,4-pentanedionato palladium ii |
| PubChem CID | 53384484 |
| IUPAC Name | palladium(2+) bis((2Z)-4-oxopent-2-en-2-olate) |
| SMILES | [Pd++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O |
Chemical Identifiers
| 14024-61-4 | |
| 304.64 | |
| JKDRQYIYVJVOPF-FDGPNNRMSA-L | |
| 53384484 | |
| [Pd++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O |
| C10H14O4Pd | |
| MFCD00000025 MFCD00000025 | |
| palladium diacetonate, acetylacetone palladium ii salt, bis 2,4-pentanedionato palladium ii | |
| palladium(2+) bis((2Z)-4-oxopent-2-en-2-olate) |
Specifications
| 14024-61-4 | |
| MFCD00000025 MFCD00000025 | |
| palladium diacetonate, acetylacetone palladium ii salt, bis 2,4-pentanedionato palladium ii | |
| [Pd++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O | |
| 304.64 | |
| 304.64 | |
| Crystalline Powder |
| C10H14O4Pd | |
| 1 g | |
| JKDRQYIYVJVOPF-FDGPNNRMSA-L | |
| palladium(2+) bis((2Z)-4-oxopent-2-en-2-olate) | |
| 53384484 | |
| ≥98.0% (T) | |
| Bis(2,4-pentanedionato)palladium(II) |
Safety and Handling
EINECSNumber : (2)-2181
TSCA : Yes