missing translation for 'onlineSavingsMsg'
Learn More

Bis(2,4-pentanedionato)palladium(II) 98.0+%, TCI America™

Catalog No. B20185G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20185G 5 g
B20181G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B20185G Supplier TCI America Supplier No. B20185G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14024-61-4
Molecular Formula C10H14O4Pd
Molecular Weight (g/mol) 304.64
MDL Number MFCD00000025 MFCD00000025
InChI Key JKDRQYIYVJVOPF-FDGPNNRMSA-L
Synonym palladium diacetonate, acetylacetone palladium ii salt, bis 2,4-pentanedionato palladium ii
PubChem CID 53384484
IUPAC Name palladium(2+) bis((2Z)-4-oxopent-2-en-2-olate)
SMILES [Pd++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Quantity 5 g
Formula Weight 304.64
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(2,4-pentanedionato)palladium(II)
EINECSNumber (2)-2181
TSCA Yes