missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(2,4-pentanedionato)molybdenum(VI) Dioxide 95.0+%, TCI America™
$95.06 - $417.56
Chemical Identifiers
| CAS | 17524-05-9 |
|---|---|
| Molecular Formula | C10H14MoO6 |
| Molecular Weight (g/mol) | 326.17 |
| MDL Number | MFCD00011506 |
| InChI Key | SKMUJBBRXZPAJY-VGKOASNMSA-L |
| Synonym | Acetylacetone Molybdenum(VI)dioxy Salt, Bis(acetylacetonato)dioxomolybdenum(VI), Molybdenum(VI)dioxy Acetylacetonate |
| IUPAC Name | dioxomolybdenumbis(ylium) bis((2Z)-4-oxopent-2-en-2-olate) |
| SMILES | O=[Mo++]=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O |
Chemical Identifiers
| 17524-05-9 | |
| 326.17 | |
| SKMUJBBRXZPAJY-VGKOASNMSA-L | |
| dioxomolybdenumbis(ylium) bis((2Z)-4-oxopent-2-en-2-olate) |
| C10H14MoO6 | |
| MFCD00011506 | |
| Acetylacetone Molybdenum(VI)dioxy Salt, Bis(acetylacetonato)dioxomolybdenum(VI), Molybdenum(VI)dioxy Acetylacetonate | |
| O=[Mo++]=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O |
Specifications
| 17524-05-9 | |
| 5 g | |
| MFCD00011506 | |
| SKMUJBBRXZPAJY-VGKOASNMSA-L | |
| dioxomolybdenumbis(ylium) bis((2Z)-4-oxopent-2-en-2-olate) | |
| 326.18 | |
| Bis(2,4-pentanedionato)molybdenum(VI) Dioxide |
| Crystalline Powder | |
| C10H14MoO6 | |
| Acetylacetone Molybdenum(VI)dioxy Salt, Bis(acetylacetonato)dioxomolybdenum(VI), Molybdenum(VI)dioxy Acetylacetonate | |
| O=[Mo++]=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O | |
| 326.17 | |
| ≥95.0% (T) |
Safety and Handling
EINECSNumber : (2)-2181
TSCA : No