missing translation for 'onlineSavingsMsg'
Learn More

Bis(2,4-pentanedionato)cobalt(II) Dihydrate 98.0+%, TCI America™

$115.51 - $716.16

Chemical Identifiers

CAS 123334-29-2
Molecular Formula C10H18CoO5
Molecular Weight (g/mol) 277.182
MDL Number MFCD00000014
InChI Key JHWSVOFBMAXGJH-SUKNRPLKSA-N
Synonym Acetylacetone Cobalt(II) Salt, Cobalt(II) Acetylacetonate
PubChem CID 22836398
IUPAC Name cobalt;(Z)-4-hydroxypent-3-en-2-one;hydrate
SMILES CC(=CC(=O)C)O.CC(=CC(=O)C)O.O.[Co]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C037325G
View Documents
TCI America
C037325G
25 g
Each for $115.51
Only null left
 
C0373500G
View Documents
TCI America
C0373500G
500 g
Each for $716.16
Only null left
 
Description

Description

Specifications

Chemical Identifiers

123334-29-2
277.182
JHWSVOFBMAXGJH-SUKNRPLKSA-N
22836398
CC(=CC(=O)C)O.CC(=CC(=O)C)O.O.[Co]
C10H18CoO5
MFCD00000014
Acetylacetone Cobalt(II) Salt, Cobalt(II) Acetylacetonate
cobalt;(Z)-4-hydroxypent-3-en-2-one;hydrate

Specifications

123334-29-2
MFCD00000014
Acetylacetone Cobalt(II) Salt, Cobalt(II) Acetylacetonate
CC(=CC(=O)C)O.CC(=CC(=O)C)O.O.[Co]
277.182
257.15
Crystalline Powder
C10H18CoO5
25 g
JHWSVOFBMAXGJH-SUKNRPLKSA-N
cobalt;(Z)-4-hydroxypent-3-en-2-one;hydrate
22836398
≥98.0% (T)
Bis(2,4-pentanedionato)cobalt(II) Dihydrate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2142&(2)-2181

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'