missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(2,4-pentanedionato)cobalt(II) Dihydrate 98.0+%, TCI America™
Specifications
Specifications
| Quantity | 25 g |
| Formula Weight | 257.15 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Bis(2,4-pentanedionato)cobalt(II) Dihydrate |
Safety and Handling
| EINECSNumber | (2)-2142&(2)-2181 |
| TSCA | Yes |