missing translation for 'onlineSavingsMsg'
Learn More

Bis(2,4,6-trimethylpyridine)bromonium Hexafluorophosphate 98.0+%, TCI America™

Catalog No. B23585G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B23585G 5 g
B23581G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B23585G Supplier TCI America Supplier No. B23585G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 188944-77-6
Molecular Formula C16H22BrF6N2P
Molecular Weight (g/mol) 467.234
InChI Key YTYGAGSIUXQVJI-UHFFFAOYSA-N
PubChem CID 44629787
IUPAC Name 2,4,6-trimethyl-1-(2,4,6-trimethylpyridin-1-ium-1-yl)bromanuidylpyridin-1-ium;hexafluorophosphate
SMILES CC1=CC(=[N+](C(=C1)C)[Br-][N+]2=C(C=C(C=C2C)C)C)C.F[P-](F)(F)(F)(F)F

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 467.23
Percent Purity ≥98.0% (HPLC)
Grade Reagent
Physical Form Crystalline Powder
Chemical Name or Material Bis(2,4,6-trimethylpyridine)bromonium Hexafluorophosphate
TSCA No
Recommended Storage Refrigerator