missing translation for 'onlineSavingsMsg'
Learn More

Bis(2,2,6,6-tetramethyl-4-piperidyl-1-oxyl) Sebacate 98.0+%, TCI America™

Catalog No. p-7124148
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B56421G 1 g
B56425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B56421G Supplier TCI America Supplier No. B56421G

Chemical Identifiers

CAS 2516-92-9
Molecular Formula C28H50N2O6
Molecular Weight (g/mol) 510.716
MDL Number MFCD12756000
InChI Key XVJWWMHPPKXKRU-UHFFFAOYSA-N
Synonym Sebacic Acid Bis(2,2,6,6-tetramethyl-4-piperidyl-1-oxyl) Ester, Bis(2,2,6,6-tetramethyl-4-piperidyl-1-oxyl) Decanedioate, Decanedioic Acid Bis(2,2,6,6-tetramethyl-4-piperidyl-1-oxyl) Ester
PubChem CID 6451408
IUPAC Name bis(1-$l^{1}-oxidanyl-2,2,6,6-tetramethylpiperidin-4-yl) decanedioate
SMILES CC1(CC(CC(N1[O])(C)C)OC(=O)CCCCCCCCC(=O)OC2CC(N(C(C2)(C)C)[O])(C)C)C

Specifications

Melting Point 101°C
Quantity 1 g
Formula Weight 510.72
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(2,2,6,6-tetramethyl-4-piperidyl-1-oxyl) Sebacate
TSCA Yes
Recommended Storage Refrigerator