missing translation for 'onlineSavingsMsg'
Learn More

Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)calcium(II), 99.99% (metals basis)

Catalog No. p-7051582
Change view
Click to view available options
Quantity:
2 g
10 g
Catalog No. Quantity
AA4158904 2 g
AA4158909 10 g
2 options

Catalog No. AA4158904

Supplier: Thermo Scientific Chemicals 04158904

Only null left

CAS: 118448-18-3 | C22H38CaO4 | 406.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 118448-18-3
Molecular Formula C22H38CaO4
Molecular Weight (g/mol) 406.62
MDL Number MFCD00064757
InChI Key DOOFPPIHJGRIGW-OLHSYEKISA-L
Synonym ca tmhd 2, calcium bis dipivaloylmethanate, bis 2,2,6,6-tetramethyl-3,5-heptanedionato calcium
PubChem CID 14598209
IUPAC Name calcium;(Z)-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate;(E)-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate
SMILES CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].[Ca+2]

Specifications

Melting Point 202°C to 206°C
Boiling Point 205°C (0.1 mmHg)(sublimation)
Quantity 2 g
Sensitivity Hygroscopic
Formula Weight 406.57
Percent Purity 99.99%
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)calcium(II)
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only