missing translation for 'onlineSavingsMsg'
Learn More

Bis(2,2,2-trifluoroethyl) Phosphite 94.0+%, TCI America™

Catalog No. p-7119699
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
B26275G 5 g
B262725G 25 g
2 options

Catalog No. B26275G

Supplier: TCI America B26275G

Only null left

Chemical Identifiers

CAS 92466-70-1
Molecular Formula C4H4F6O3P+
Molecular Weight (g/mol) 245.037
InChI Key IMDCVAFSSZPRRM-UHFFFAOYSA-N
Synonym bis 2,2,2-trifluoroethyl phosphonate, phosphonic acid bis 2,2,2-trifluoroethyl ester, oxo-bis 2,2,2-trifluoroethoxy phosphanium, phosphonic acid, bis 2,2,2-trifluoroethyl ester, bis trifluoroethyl phosphonate, bis 2,2,2-trifluoroethyl phosphite, bis 2,2,2-trifluoroethyl hydrogen phosphite, oxobis 2,2,2-trifluoroethoxy-??-phosphanylium, bis 2,2,2-trifluoroethyl phosphite, technical grade
PubChem CID 6386648
IUPAC Name oxo-bis(2,2,2-trifluoroethoxy)phosphanium
SMILES C(C(F)(F)F)O[P+](=O)OCC(F)(F)F

Specifications

Color Yellow
Boiling Point 44°C
Quantity 5 g
Formula Weight 246.05
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material Bis(2,2,2-trifluoroethyl) Phosphite