missing translation for 'onlineSavingsMsg'
Learn More

Bis(1,4-phenylene)-34-crown 10-Ether, TCI America™

Catalog No. b2245100mg
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2245100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B2245100MG Supplier TCI America Supplier No. B2245100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 53914-95-7
Molecular Formula C28H40O10
Molecular Weight (g/mol) 536.618
MDL Number MFCD03846057
InChI Key REKDBTBSNFSNGP-UHFFFAOYSA-N
PubChem CID 635862
SMILES C1COCCOC2=CC=C(C=C2)OCCOCCOCCOCCOC3=CC=C(C=C3)OCCOCCO1

Specifications

Color White
Quantity 100 mg
Formula Weight 536.62
Physical Form Crystalline Powder
Chemical Name or Material Bis(1,4-phenylene)-34-crown 10-Ether