missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(1,3-propanediamine) Copper(II) Dichloride 98.0+%, TCI America™
$68.97 - $417.56
Chemical Identifiers
| CAS | 32270-93-2 |
|---|---|
| Molecular Formula | C6H20Cl2CuN4 |
| Molecular Weight (g/mol) | 282.7 |
| MDL Number | MFCD00191318 |
| InChI Key | HSUSKWQWIPIFEB-UHFFFAOYSA-L |
| PubChem CID | 12980278 |
| IUPAC Name | dichlorocopper;propane-1,3-diamine |
| SMILES | C(CN)CN.C(CN)CN.Cl[Cu]Cl |
Chemical Identifiers
| 32270-93-2 | |
| 282.7 | |
| HSUSKWQWIPIFEB-UHFFFAOYSA-L | |
| dichlorocopper;propane-1,3-diamine |
| C6H20Cl2CuN4 | |
| MFCD00191318 | |
| 12980278 | |
| C(CN)CN.C(CN)CN.Cl[Cu]Cl |
Specifications
| 32270-93-2 | |
| C6H20Cl2CuN4 | |
| 1 g | |
| C(CN)CN.C(CN)CN.Cl[Cu]Cl | |
| 282.7 | |
| 282.70 | |
| Crystalline Powder |
| Blue-Purple | |
| MFCD00191318 | |
| HSUSKWQWIPIFEB-UHFFFAOYSA-L | |
| dichlorocopper;propane-1,3-diamine | |
| 12980278 | |
| ≥98.0% (T) | |
| Bis(1,3-propanediamine) Copper(II) Dichloride |
Safety and Handling
TSCA : No