missing translation for 'onlineSavingsMsg'
Learn More

Bis[1,2-bis(diphenylphosphino)ethane]palladium(0) 96.0+%, TCI America™

Catalog No. p-7122818
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
B32241G 1 g
B32245G 5 g
2 options

Catalog No. B32241G

Supplier: TCI America B32241G

Only null left

Chemical Identifiers

CAS 31277-98-2
Molecular Formula C52H48P4Pd
Molecular Weight (g/mol) 903.27
MDL Number MFCD00009880
InChI Key FAFGMAGIYHHRKN-UHFFFAOYSA-N
Synonym bis 1,2-bis diphenylphosphino ethane palladium 0, pd dppe 2, pd diphos 2, bis diphos palladium 0, bis 1,2-bis diphenylphosphino ethane palladium, bis diphos palladium, bis 1,2-bis diphenylphosphino ethane-palladium 0, bis 1,2-bis diphenylphosphino ethane-palladium
PubChem CID 2724231
IUPAC Name bis([2-(diphenylphosphanyl)ethyl]diphenylphosphane) palladium
SMILES [Pd].C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Quantity 1 g
Formula Weight 903.27
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis[1,2-bis(diphenylphosphino)ethane]palladium(0)
TSCA No
Recommended Storage Freezer (-20 °C)