missing translation for 'onlineSavingsMsg'
Learn More

Bis[1,2-bis(diphenylphosphino)ethane]palladium(0) 96.0+%, TCI America™

Catalog No. B32241G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32241G 1 g
B32245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B32241G Supplier TCI America Supplier No. B32241G

Chemical Identifiers

CAS 31277-98-2
Molecular Formula C52H48P4Pd
Molecular Weight (g/mol) 903.27
MDL Number MFCD00009880
InChI Key FAFGMAGIYHHRKN-UHFFFAOYSA-N
Synonym bis 1,2-bis diphenylphosphino ethane palladium 0, pd dppe 2, pd diphos 2, bis diphos palladium 0, bis 1,2-bis diphenylphosphino ethane palladium, bis diphos palladium, bis 1,2-bis diphenylphosphino ethane-palladium 0, bis 1,2-bis diphenylphosphino ethane-palladium
PubChem CID 2724231
IUPAC Name bis([2-(diphenylphosphanyl)ethyl]diphenylphosphane) palladium
SMILES [Pd].C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Quantity 1 g
Formula Weight 903.27
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis[1,2-bis(diphenylphosphino)ethane]palladium(0)
TSCA No
Recommended Storage Freezer (-20 °C)