missing translation for 'onlineSavingsMsg'
Learn More

Bis[1,2,3,4,5-pentaphenyl-1'-(di-tert-butylphosphino)ferrocene]palladium(0), Thermo Scientific Chemicals

Catalog No. p-7059998
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4591303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4591303 Supplier Thermo Scientific Chemicals Supplier No. 04591303

Bis[1,2,3,4,5-pentaphenyl-1'-(di-tert-butylphosphino)ferrocene]palladium(0) is used to prepare palladium(0) catalysts for highly selective cross-coupling reactions especially carbohalogenation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 565441-56-7
Molecular Formula C96H94Fe2P2Pd
Molecular Weight (g/mol) 1527.87
MDL Number MFCD15071401
Synonym Pd(Qhos)2
IUPAC Name Bis[1,2,3,4,5-pentaphenyl-1'-(di-tert-butylphosphino)ferrocene]palladium(0)
SMILES [Fe].[Fe].[Pd].CC(C)(C)P(c1cccc1)C(C)(C)C.CC(C)(C)P(c1cccc1)C(C)(C)C.C1=CC=C(C=C1)c1c(c(c(c1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)c1c(c(c(c1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Pink to White
Quantity 1 g
Sensitivity Air sensitive
Solubility Information Insoluble in water.
Formula Weight 1527.86
Physical Form Crystalline
Chemical Name or Material Bis[1,2,3,4,5-pentaphenyl-1'- (di-tert-butylphosphino)ferrocene]palladium (0)
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only