missing translation for 'onlineSavingsMsg'
Learn More

Biphenylene, 99+%, Thermo Scientific Chemicals

Catalog No. AC279742500
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC279742500 250 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC279742500 Supplier Thermo Scientific Chemicals Supplier No. 279742500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 259-79-0 | Molecular Formula: C12H8 | Molecular Weight: 152.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 259-79-0
Molecular Formula C12H8
Molecular Weight (g/mol) 152.2
MDL Number MFCD00001110
InChI Key IFVTZJHWGZSXFD-UHFFFAOYSA-N
Synonym diphenylene, cyclobutadibenzene, dibenzocyclobutadiene, 1,1'-biphenylene, unii-0z64i7d5m2, 8ci 9ci, biphenylen, biphenylene, biphenylene, 99+%, pubchem9106
PubChem CID 9214
ChEBI CHEBI:33079
IUPAC Name biphenylene
SMILES C1=CC=C2C(=C1)C3=CC=CC=C23

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 109°C to 112°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 250 mg
Beilstein 05, 298
Solubility Information Solubility in water: 9.60mg/l (calc). Other solubilities: organic solvents general
Formula Weight 152.2
Percent Purity 99+%
Physical Form Needles
Chemical Name or Material Biphenylene, 99+%
Show More Show Less
Hazard Statement GHS H Statement:
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only