missing translation for 'onlineSavingsMsg'
Learn More

Biphenyl-4-carboxylic Hydrazide 97.0+%, TCI America™

Catalog No. B43501G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B43501G Supplier TCI America Supplier No. B43501G
Only null left

Chemical Identifiers

CAS 18622-23-6
Molecular Formula C13H12N2O
Molecular Weight (g/mol) 212.252
MDL Number MFCD00017078
InChI Key QEUAQXSDDNDOTG-UHFFFAOYSA-N
Synonym 4-Phenylbenzoic Hydrazide, 4-Phenylbenzoylhydrazine
PubChem CID 72718
IUPAC Name 4-phenylbenzohydrazide
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)NN

Specifications

Melting Point 195°C
Color White
Quantity 1 g
Formula Weight 212.25
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Biphenyl-4-carboxylic Hydrazide