missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biphenyl-4-carboxaldehyde 95.0+%, TCI America™
$55.84 - $779.69
Chemical Identifiers
| CAS | 3218-36-8 |
|---|---|
| Molecular Formula | C13H10O |
| Molecular Weight (g/mol) | 182.22 |
| MDL Number | MFCD00006947 |
| InChI Key | ISDBWOPVZKNQDW-UHFFFAOYSA-N |
| Synonym | 4-biphenylcarboxaldehyde, 4-biphenylaldehyde, p-phenylbenzaldehyde, 1,1'-biphenyl-4-carbaldehyde, biphenyl-4-carboxaldehyde, 4-formylbiphenyl, p-biphenylylaldehyde, 1,1'-biphenyl-4-carboxaldehyde, p-biphenylaldehyde, p-biphenylcarboxaldehyde |
| PubChem CID | 76689 |
| IUPAC Name | [1,1'-biphenyl]-4-carbaldehyde |
| SMILES | O=CC1=CC=C(C=C1)C1=CC=CC=C1 |
Chemical Identifiers
| 3218-36-8 | |
| 182.22 | |
| ISDBWOPVZKNQDW-UHFFFAOYSA-N | |
| 76689 | |
| O=CC1=CC=C(C=C1)C1=CC=CC=C1 |
| C13H10O | |
| MFCD00006947 | |
| 4-biphenylcarboxaldehyde, 4-biphenylaldehyde, p-phenylbenzaldehyde, 1,1'-biphenyl-4-carbaldehyde, biphenyl-4-carboxaldehyde, 4-formylbiphenyl, p-biphenylylaldehyde, 1,1'-biphenyl-4-carboxaldehyde, p-biphenylaldehyde, p-biphenylcarboxaldehyde | |
| [1,1'-biphenyl]-4-carbaldehyde |
Specifications
| 3218-36-8 | |
| 152°C | |
| MFCD00006947 | |
| 4-biphenylcarboxaldehyde, 4-biphenylaldehyde, p-phenylbenzaldehyde, 1,1'-biphenyl-4-carbaldehyde, biphenyl-4-carboxaldehyde, 4-formylbiphenyl, p-biphenylylaldehyde, 1,1'-biphenyl-4-carboxaldehyde, p-biphenylaldehyde, p-biphenylcarboxaldehyde | |
| O=CC1=CC=C(C=C1)C1=CC=CC=C1 | |
| 182.22 | |
| 182.22 | |
| Crystalline Powder |
| 58°C | |
| C13H10O | |
| 5 g | |
| ISDBWOPVZKNQDW-UHFFFAOYSA-N | |
| [1,1'-biphenyl]-4-carbaldehyde | |
| 76689 | |
| ≥95.0% (GC) | |
| Biphenyl-4-carboxaldehyde |
Safety and Handling
EINECSNumber : (4)-1745
RTECSNumber : DV1767000
TSCA : No