missing translation for 'onlineSavingsMsg'
Learn More

Biphenyl-4-carboxaldehyde, 98+%

Catalog No. AAA1583906
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1583906 5 g
AAA1583914 25 g
AAA1583922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1583906 Supplier Thermo Scientific Chemicals Supplier No. A1583906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3218-36-8 | C13H10O | 182.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3218-36-8
Molecular Formula C13H10O
Molecular Weight (g/mol) 182.22
MDL Number MFCD00006947
InChI Key ISDBWOPVZKNQDW-UHFFFAOYSA-N
Synonym 4-biphenylcarboxaldehyde, 4-biphenylaldehyde, p-phenylbenzaldehyde, 1,1'-biphenyl-4-carbaldehyde, biphenyl-4-carboxaldehyde, 4-formylbiphenyl, p-biphenylylaldehyde, 1,1'-biphenyl-4-carboxaldehyde, p-biphenylaldehyde, p-biphenylcarboxaldehyde
PubChem CID 76689
IUPAC Name 4-phenylbenzaldehyde
SMILES O=CC1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 57°C to 61°C
Density 1.107
Boiling Point 184°C (11 mmHg)
Flash Point 172°C (341°F)
Quantity 5 g
Beilstein 606693
Sensitivity Air Sensitive
Formula Weight 182.22
Percent Purity ≥98%
Chemical Name or Material Biphenyl-4-carboxaldehyde
Hazard Category H302
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 221-742-3
RTECSNumber DV1767000
TSCA Yes

RUO – Research Use Only