missing translation for 'onlineSavingsMsg'
Learn More

Biphenyl-4-boronic acid pinacol ester, 97%

Catalog No. p-7040210
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5991903 1 g
AAH5991906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5991903 Supplier Thermo Scientific Chemicals Supplier No. H5991903

CAS: 144432-80-4 | C18H21BO2 | 280.17 g/mol

It is an important raw material and intermediate used in organic synthesis and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis and pharmaceuticals.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 144432-80-4
Molecular Formula C18H21BO2
Molecular Weight (g/mol) 280.17
MDL Number MFCD07368291
InChI Key REDKQKNJWVIPIO-UHFFFAOYSA-N
Synonym 4-biphenylboronic acid pinacol ester, 2-1,1'-biphenyl-4-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-4-biphenylyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-biphenylboronic acid, pinacol ester, 4,4,5,5-tetramethyl-2-4-phenylphenyl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 2-1,1'-biphenyl-4-yl-4,4,5,5-tetramethyl, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl biphenyl, 2-biphenyl-4-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl diphenyl
PubChem CID 11989651
IUPAC Name 4,4,5,5-tetramethyl-2-(4-phenylphenyl)-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 280.17
Percent Purity 97%
Chemical Name or Material Biphenyl-4-boronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only