missing translation for 'onlineSavingsMsg'
Learn More

Biphenyl-4-boronic acid, 97+%

Catalog No. AAB2370303
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2370303 1 g
AAB2370306 5 g
AAB2370314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2370303 Supplier Thermo Scientific Chemicals Supplier No. B2370303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5122-94-1 | C12H11BO2 | 198.03 g/mol

Used in Suzuki reactions and as intermediates of liquid crystals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in Suzuki reactions and as intermediates of liquid crystals.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 5122-94-1
Molecular Formula C12H11BO2
Molecular Weight (g/mol) 198.03
MDL Number MFCD00093311
InChI Key XPEIJWZLPWNNOK-UHFFFAOYSA-N
Synonym 4-biphenylboronic acid, 1,1'-biphenyl-4-ylboronic acid, biphenyl-4-boronic acid, 4-biphenyl boronic acid, biphenylboronic acid, 4-phenylphenyl boranediol, 4-phenylphenyl boronic acid, biphenyl-4-ylboronic acid, 1,1'-biphenyl-4-yl-boronic acid, 4-phenylbenzeneboronic acid
PubChem CID 151253
IUPAC Name (4-phenylphenyl)boronic acid
SMILES OB(O)C1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 238°C to 247°C
Odor Odorless
Quantity 1 g
Beilstein 2937410
Solubility Information Insoluble in water.
Formula Weight 198.03
Percent Purity ≥98%
Chemical Name or Material Biphenyl-4-boronic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only