missing translation for 'onlineSavingsMsg'
Learn More

Biphenyl-4,4'-diboronic acid bis(pinacol) ester, 95%

Catalog No. AAH5195606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5195606 5 g
AAH5195603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5195606 Supplier Thermo Scientific Chemicals Supplier No. H5195606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 207611-87-8 | C24H32B2O4 | 406.14 g/mol

Biphenyl-4,4'-diboronic acid bis(pinacol) ester is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Biphenyl-4,4′-diboronic acid bis(pinacol) ester is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 207611-87-8
Molecular Formula C24H32B2O4
Molecular Weight (g/mol) 406.14
MDL Number MFCD08669545
InChI Key INNVTCAYPFZCAL-UHFFFAOYSA-N
Synonym 4,4'-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1,1'-biphenyl, 4,4'-biphenyldiboronic acid dipinacol ester, 4,4'-biphenyldiboronic acid pinacol ester, 4,4'-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl biphenyl, biphenyl-4,4'-diboronic acid bis pinacol ester, 4,4'-biphenyldiboronic acid bis pinacol ester, 4,4,5,5-tetramethyl-2-4'-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1,1'-biphenyl-4-yl-1,3,2-dioxaborolane, amtb257, 4,4'-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl biphenyl
PubChem CID 10525359
IUPAC Name 4,4,5,5-tetramethyl-2-[4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]phenyl]-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Specifications

Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 406.14
Percent Purity 95%
Chemical Name or Material Biphenyl-4,4'-diboronic acid bis(pinacol) ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only