missing translation for 'onlineSavingsMsg'
Learn More

Biphenyl-3-carboxaldehyde 96.0+%, TCI America™

Catalog No. p-7123396
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41861G 1 g
B41865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B41861G Supplier TCI America Supplier No. B41861G

Chemical Identifiers

CAS 1204-60-0
Molecular Formula C13H10O
Molecular Weight (g/mol) 182.222
MDL Number MFCD01740432
InChI Key KFKSIUOALVIACE-UHFFFAOYSA-N
Synonym 1,1'-biphenyl-3-carbaldehyde, biphenyl-3-carbaldehyde, 3-formylbiphenyl, ccris 3164, biphenyl-3-carboxaldehyde, 3-difenilaldeide italian, 1,1'-biphenyl-3-carboxaldehyde, 3-biphenyl carboxaldehyde
PubChem CID 121053
IUPAC Name 3-phenylbenzaldehyde
SMILES C1=CC=C(C=C1)C2=CC(=CC=C2)C=O

Specifications

Color Yellow
Boiling Point 125°C
Quantity 1 g
Formula Weight 182.22
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Biphenyl-3-carboxaldehyde
RTECSNumber DV1771500
TSCA No