missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biphenyl-2,2'-dicarboxaldehyde 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 64°C |
| Color | Yellow |
| Boiling Point | 179°C |
| Quantity | 100 mg |
| Formula Weight | 210.23 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Biphenyl-2,2′-dicarboxaldehyde |
Safety and Handling
| RTECSNumber | JJ3850000 |
| TSCA | No |