missing translation for 'onlineSavingsMsg'
Learn More

BioTracker 543 Yellow β-Gal Dye, MilliporeSigma™

Catalog No. SCT024
Change view
Click to view available options
Quantity:
10 x 50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCT024 10 x 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCT024 Supplier MilliporeSigma Supplier No. SCT024

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Live cell imaging dye for β-galactosidase (β-gal) used to detect LacZ reporter gene expression and cellular senescence (SA-β-gal).

  • β-galactosidase, also called beta-gal or β-gal, is a glycoside hydrolase enzyme that catalyzes the hydrolysis of β-galactosides into monosaccharides through the breaking of a glycosidic bond. β-gal is commonly used in molecular biology as a reporter marker to monitor gene expression using the chromogenic X-Gal. β-gal has also been used to measure cellular senescence (SA-β-gal).
  • The BioTracker 543 Yellow β-Gal Dye is a fluorescent probe for the detection of β-galactosidase in living cells. It can be applied to fluorescent imaging and selection of cell and tissue transfected with lacZ along as a measurement of cellular senescence. Other applications include gene analysis by fluorescent imaging, monitoring transfection efficiency, and the study of gene promoter or enhancer elements. Because the dye is almost non-fluorescent in the absence of β-galactosidase, this probe exhibits good S/N ratio.
  • Absorbance: 525 nm (488 nm compatible)
  • Emission: 543 nm

Specifications

Absorbance 525 nm
CAS 1326231-95-1
Molecular Weight (g/mol) 535.58
Grade Research
Quantity 10 x 50 μg