Learn More
Eisco™ Biotite Mica Mineral Specimen
Biotite Mica Specimen suitable for use in classroom identification lab exercises and for study on 2-plane, 90° cleavage.
$10.25 - $90.64
Specifications
Product Type Mineral, Biotite Mica
Specimen Size 2 to 3 cm
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
S24675
View Documents Promotion Details
Eisco™
ESNG0012
1 Ea.
Each for $10.25
Only null left
S26616
View Documents Promotion Details
Eisco™
ESNG0012PK10
10/Pk.
Each for $90.64
Only null left
Description
Description
  • Pair with muscovite mica to demonstrate effects of slight variations in a mineral's chemistry
  • Useful when teaching lessons about physical properties of minerals, as well as Bowen's reaction series. Couple samples can also act as polarizers
  • Useful when teaching students about variable hardness and cleavage
  • Mica's thinly layered structure is unique
  • Correlates well to Next Generation Science Standards Evidence Statements
Specifications
Specifications
Mineral, Biotite Mica
2 to 3 cm