Learn More
ACROBiosystems Inc Biotinylated SARS-CoV-2 (COVID-19) Nucleocapsid protein, His,Avitag™
Catalog No. 502055148
Encompass
Recombinant Protein;200UG;Biotinylated SARS-CoV-2 Nucleocapsid protein, His,Avitag™ (NUN-C82E8) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 419 (Accession # QHO62115.1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 49.0 kDa. The protein migrates as 55-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-SARS-CoV-2 Nucleocapsid Antibody, Human IgG1 (Cat. No. NUN-S41) at 1 g/mL (100 L/well) can bind Biotinylated SARS-CoV-2 Nucleocapsid protein, His,Avitag (Cat. No. NUN-C82E8) with a linear range of 0.2-6 ng/mL (QC tested).
Catalog No. 502055148 Supplier ACROBiosystems Inc Supplier No. NUNC82E8200UG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant Protein;200UG;Biotinylated SARS-CoV-2 Nucleocapsid protein, His,Avitag™ (NUN-C82E8) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 419 (Accession # QHO62115.1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 49.0 kDa. The protein migrates as 55-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-SARS-CoV-2 Nucleocapsid Antibody, Human IgG1 (Cat. No. NUN-S41) at 1 g/mL (100 L/well) can bind Biotinylated SARS-CoV-2 Nucleocapsid protein, His,Avitag (Cat. No. NUN-C82E8) with a linear range of 0.2-6 ng/mL (QC tested).