Learn More
ACROBiosystems Inc Biotinylated Human PSCA Protein, His,Avitag™
Catalog No. 502054760
Encompass
Recombinant Protein;200UG;Biotinylated Human PSCA, His,Avitag (PSA-H82E3) is expressed from human 293 cells (HEK293). It contains AA Leu 12 - Ser 86 (Accession # O43653-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 12.1 kDa.
Catalog No. 502054760 Supplier ACROBiosystems Inc Supplier No. PSAH82E3200UG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant Protein;200UG;Biotinylated Human PSCA, His,Avitag (PSA-H82E3) is expressed from human 293 cells (HEK293). It contains AA Leu 12 - Ser 86 (Accession # O43653-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 12.1 kDa.