Recombinant Protein;200UG;Biotinylated Human M-CSF R, Avitag,His Tag (CSR-H82E0) is expressed from human 293 cells (HEK293). It contains AA Ile 20 - Glu 512 (Accession # P07333-1).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 57.1 kDa. The protein migrates as 65-92 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human M-CSF, His Tag (Cat. No. MCF-H5247) at 5 g/mL (100 L/well) can bind Biotinylated Human M-CSF R, Avitag,His Tag (Cat. No. CSR-H82E0) with a linear range of 16-500 ng/mL (QC tested).