Recombinant Protein;25UG;Biotinylated Human CD45, His,Avitag (CD5-H82E7) is expressed from human 293 cells (HEK293). It contains AA Gln 26-Lys 577 (Accession # P08575-3).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 64.3 kDa.