Learn More
ACROBiosystems Inc ACROBiosystems Biotinylated Human CD4 Protein, His,Avitag™ (MALS verified)
Catalog No. 502106555
Encompass
Recombinant Protein;200UG;Biotinylated Human CD4, His,Avitag (CD4-H82E8) is expressed from human 293 cells (HEK293). It contains AA Lys 26 - Pro 396 (Accession # P01730-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 45.0 kDa. The protein migrates as 55-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Ibalizumab at 1 g/mL (100 L/well) can bind Biotinylated Human CD4, His,Avitag (Cat. No. CD4-H82E8) with a linear range of 0.2-3 ng/mL (QC tested).;Immobilized HIV-1 [HIV-1/Clade C (16055)] GP120, His Tag (Cat. No. GP5-V15224) at 5 g/mL (100 L/well) can bind Biotinylated Human CD4, His,Avitag (Cat. No. CD4-H82E8) with a linear range of 1-16 ng/mL (Routinely tested).
Catalog No. 502106555 Supplier ACROBiosystems Inc Supplier No. CD4H82E8200UG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant Protein;200UG;Biotinylated Human CD4, His,Avitag (CD4-H82E8) is expressed from human 293 cells (HEK293). It contains AA Lys 26 - Pro 396 (Accession # P01730-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 45.0 kDa. The protein migrates as 55-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Ibalizumab at 1 g/mL (100 L/well) can bind Biotinylated Human CD4, His,Avitag (Cat. No. CD4-H82E8) with a linear range of 0.2-3 ng/mL (QC tested).;Immobilized HIV-1 [HIV-1/Clade C (16055)] GP120, His Tag (Cat. No. GP5-V15224) at 5 g/mL (100 L/well) can bind Biotinylated Human CD4, His,Avitag (Cat. No. CD4-H82E8) with a linear range of 1-16 ng/mL (Routinely tested).