Recombinant Protein;25UG;Biotinylated Human CD2, His,Avitag (CD2-H82E6) is expressed from human 293 cells (HEK293). It contains AA Lys 25 - Asp 209 (Accession # AAH33583).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 24.8 kDa. The protein migrates as 35-43 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.;Immobilized Human CD58, Fc Tag (Cat. No. LF3-H5256) at 5 g/mL (100 L/well) can bind Biotinylated Human CD2, His,Avitag (Cat. No. CD2-H82E6) with a linear range of 0.2-1.25 g/mL (QC tested).