Learn More
ACROBiosystems Inc Recombinant Protein;Biotinylated Human VEGF165, epitope tag free, ultra sensitivity (primary amine labeling);HEK293;200UG;VE5,RP1-261G23.1,MGC70609,MVCD1,MVCD-1,VEGF 165,VEGF-A,VEGFA,VPF,VEGF165,VEGF-165,VEGF121,VEGF120,VEGF186,VEGF164,VEGF110
Catalog No. 501018749
Encompass
Recombinant Protein;Biotinylated Human VEGF165, epitope tag free, ultra sensitivity (primary amine labeling);200UG;MABSol Biotinylated Human VEGF165 (VE5-H8210) is expressed from human HEK293 cells. It contains AA Ala 27 - Arg 191 (Accession # NP_001165097). It is the biotinylated form of ActiveMax Recombinant Human VEGF165 (Cat # VE5-H4210).;The product does NOT contain any epitope tags. The protein has a calculated MW of 19 kDa (monomer). The protein migrates as 24 kDa (monomer) on a SDS-PAGE gel under reducing (R) condition due to glycosylation and 43-50 kDa under non-reducing (NR) condition.;Immobilized human VEGFR1/R2, Fc tag at 2g/mL (100 L/well) can bind Biotinylated Human VEGF (Cat. No. VE5-H8210) with a linear range of 0.39-3.1 ng/mL (QC tested).;Immobilized Human Neuropilin-1, His Tag (Cat.No. NR1-H5228) at 2 g/mL (100 L/well) can bind Biotinylated Human VEGF165 (Cat.No. VE5-H8210) with a linear range of 5-40 ng/mL (Routinely tested).
Catalog No. 501018749 Supplier ACROBiosystems Inc Supplier No. VE5H8210200UG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant Protein;Biotinylated Human VEGF165, epitope tag free, ultra sensitivity (primary amine labeling);200UG;MABSol Biotinylated Human VEGF165 (VE5-H8210) is expressed from human HEK293 cells. It contains AA Ala 27 - Arg 191 (Accession # NP_001165097). It is the biotinylated form of ActiveMax Recombinant Human VEGF165 (Cat # VE5-H4210).;The product does NOT contain any epitope tags. The protein has a calculated MW of 19 kDa (monomer). The protein migrates as 24 kDa (monomer) on a SDS-PAGE gel under reducing (R) condition due to glycosylation and 43-50 kDa under non-reducing (NR) condition.;Immobilized human VEGFR1/R2, Fc tag at 2g/mL (100 L/well) can bind Biotinylated Human VEGF (Cat. No. VE5-H8210) with a linear range of 0.39-3.1 ng/mL (QC tested).;Immobilized Human Neuropilin-1, His Tag (Cat.No. NR1-H5228) at 2 g/mL (100 L/well) can bind Biotinylated Human VEGF165 (Cat.No. VE5-H8210) with a linear range of 5-40 ng/mL (Routinely tested).