missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biotin, Powder, FCC, 97.5-100.5%, Spectrum™ Chemical
BI115, 58-85-5, C10H16N2O3S
Supplier: Spectrum Chemical Mfg Cor BI1155GM
Description
Spectrum™ Chemical Biotin, Powder, FCC is often recommended as a dietary supplement for strenghening hair and nails. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 58-85-5 | |
| 229-232°C | |
| C10H16N2O3S | |
| 5 g | |
| +89° to +93° | |
| 5-{2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl}pentanoic acid | |
| 97.5 to 100.5% |
| 100% | |
| Amber Glass Bottle | |
| MFCD00005541 | |
| YBJHBAHKTGYVGT-UHFFFAOYNA-N | |
| OC(=O)CCCCC1SCC2NC(=O)NC12 | |
| 244.31 | |
| FCC |
Chemical Identifiers
| 58-85-5 | |
| 244.31 | |
| YBJHBAHKTGYVGT-UHFFFAOYNA-N | |
| OC(=O)CCCCC1SCC2NC(=O)NC12 |
| C10H16N2O3S | |
| MFCD00005541 | |
| 5-{2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl}pentanoic acid |