missing translation for 'onlineSavingsMsg'
Learn More

Biotin-PEG3-Amine 95.0+%, TCI America™

Catalog No. p-7119999
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B317225MG 25 mg
B3172100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B317225MG Supplier TCI America Supplier No. B317225MG
Only null left

Chemical Identifiers

CAS 359860-27-8
Molecular Formula C18H34N4O5S
Molecular Weight (g/mol) 418.553
MDL Number MFCD09952640
InChI Key KIJSBKNJFAUJFV-ZOBUZTSGSA-N
Synonym N-Biotinyl-3,6,9-trioxaundecane-1,11-diamine, N-[2-[2-[2-(2-Aminoethoxy)ethoxy]ethoxy]ethyl]biotinamide
PubChem CID 50896259
IUPAC Name 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]-N-[2-[2-[2-(2-aminoethoxy)ethoxy]ethoxy]ethyl]pentanamide
SMILES C1C2C(C(S1)CCCCC(=O)NCCOCCOCCOCCN)NC(=O)N2

Specifications

Color White
Quantity 25 mg
Formula Weight 418.55
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Biotin-PEG3-Amine
TSCA No
Recommended Storage Freezer