missing translation for 'onlineSavingsMsg'
Learn More

Biotin-PEG11-Amine 93.0+%, TCI America™

Catalog No. p-7124119
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B556510MG 10 mg
B556550MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B556510MG Supplier TCI America Supplier No. B556510MG
Only null left

Chemical Identifiers

CAS 1418022-42-0
Molecular Formula C34H66N4O13S
Molecular Weight (g/mol) 770.977
InChI Key SHPHAOSBFNWGAC-PHDGFQFKSA-N
Synonym N-Biotinyl-3,6,9,12,15,18,21,24,27,30,33-undecaoxapentatriacontane-1,35-diamine, N-[2-[2-[2-[2-[2-[2-[2-[2-[2-[2-[2-(2-Aminoethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethyl]biotinamide
PubChem CID 77078444
IUPAC Name 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]-N-[2-[2-[2-[2-[2-[2-[2-[2-[2-[2-[2-(2-aminoethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethyl]pentanamide
SMILES C1C2C(C(S1)CCCCC(=O)NCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCN)NC(=O)N2

Specifications

Color White
Quantity 10 mg
Formula Weight 770.98
Percent Purity ≥93.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Biotin-PEG11-Amine
TSCA No
Recommended Storage Freezer